APO455834223 Apollo Scientific Ltd

2-Benzyloxy-5-fluorobenzeneboronic acid

  • MDL number MFCD03788403
  • Cas Number : 779331-47-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8641

Safety Information

signalword Warning
pcodes P280;P264;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8641
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart