APO455834228 Apollo Scientific Ltd

3-Methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazol-5-amine

  • MDL number MFCD03791147
  • Cas Number : 851975-08-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 110-113
Synonym 5-Amino-3-methyl-4-[3-(trifluoromethyl)phenyl]-1H-pyrazole, 3-(5-Amino-3-methyl-1H-pyrazol-4-yl)benzotrifluoride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=8978

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=8978
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart