APO455834231 Apollo Scientific Ltd

5-Amino-2-chloro-4-fluorophenol

  • MDL number MFCD00055622
  • Cas Number : 84478-72-8
  • Molecular Weight : 161.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 157-160
Synonym 4-Chloro-2-fluoro-5-hydroxyaniline
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-amino-2-chloro-4-fluorophenol

Safety Information

signalword Warning
pcodes P260;P280;P301+P310;P304+P340;P305+P351+P338;P312
MSDS https://store.apolloscientific.co.uk/product/5-amino-2-chloro-4-fluorophenol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart