APO455834236 Apollo Scientific Ltd

1,1,1-Trifluoropent-4-en-2-ol

  • MDL number MFCD07784256
  • Cas Number : 77342-37-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5,5,5-Trifluoropent-1-en-4-ol, 4-Hydroxy-5,5,5-trifluoropent-1-ene
Flash Point 57
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 100-101
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9582

Safety Information

signalword Warning
pcodes P210;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9582
pictograms GHS02
hstatements H226
Availability details for
Quantity
  • Availability
  • Add to Cart