APO455834237 Apollo Scientific Ltd

Ethyl 1-(tert-butyl)-5-(trifluoromethyl)-1H-pyrazole-4-carboxylate

  • MDL number MFCD04973716
  • Cas Number : 852691-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(tert-Butyl)-4-(ethoxycarbonyl)-5-(trifluoromethyl)-1H-pyrazole
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9007

Safety Information

signalword Warning
pcodes P261;P280;P271
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9007
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart