APO455834245 Apollo Scientific Ltd

5-(Chloromethyl)-4-methyl-2-[4-(trifluoromethyl)phenyl]-1,3-thiazole

  • MDL number MFCD03791173
  • Cas Number : 317318-97-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-62
Synonym 4-[5-(Chloromethyl)-4-methyl-1,3-thiazol-2-yl]benzotrifluoride
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/5-chloromethyl-4-methyl-2-4-trifluoromethylphenyl-13-thiazole

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-chloromethyl-4-methyl-2-4-trifluoromethylphenyl-13-thiazole
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart