APO455834246 Apollo Scientific Ltd

2-[3-(Trifluoromethyl)benzyl]-1,3-thiazole-4-carboxylic acid

  • MDL number MFCD02186742
  • Cas Number : 478030-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 123-125
Synonym 2-{[3-(Trifluoromethyl)phenyl]methyl}-1,3-thiazole-4-carboxylic acid, 3-[(4-Carboxy-1,3-thiazol-2-yl)methyl]benzotrifluoride
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9047

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9047
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart