APO455834247 Apollo Scientific Ltd

1-{3-[(2-Chloro-6-fluorobenzyl)oxy]phenyl}ethan-1-one

  • MDL number MFCD03001254
  • Cas Number : 400878-26-4
  • Molecular Weight : 278.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 60-62
Synonym 3'-[(2-Chloro-6-fluorobenzyl)oxy]acetophenone, 3'-[(2-Chloro-6-fluorophenyl)methoxy]acetophenone
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-3-2-chloro-6-fluorobenzyloxyphenylethan-1-one

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/1-3-2-chloro-6-fluorobenzyloxyphenylethan-1-one
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart