APO455834253 Apollo Scientific Ltd

Ethyl 2,3-dihydro-2-oxo-4-(trifluoromethyl)-1,3-thiazole-5-carboxylate

  • MDL number MFCD03425775
  • Cas Number : 72850-53-4
  • Molecular Weight : 241.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 122-124
Iupac Name ethyl 2-oxo-4-(trifluoromethyl)-2,3-dihydro-1,3-thiazole-5-carboxylate
Inchi Code 1S/C7H6F3NO3S/c1-2-14-5(12)3-4(7(8,9)10)11-6(13)15-3/h2H2,1H3,(H,11,13)
Inchi Key CTGOETARURNCLO-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 122-124
Product Link https://store.apolloscientific.co.uk/product/ethyl-23-dihydro-2-oxo-4-trifluoromethyl-13-thiazole-5-carboxylate

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-23-dihydro-2-oxo-4-trifluoromethyl-13-thiazole-5-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart