APO455834258 Apollo Scientific Ltd

2-[(2-Chloro-6-fluorobenzyl)oxy]benzaldehyde

  • MDL number MFCD01590675
  • Cas Number : 336880-01-4
  • Molecular Weight : 264.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 71-73
Synonym 2-[(2-Chloro-6-fluorophenyl)methoxy]benzaldehyde
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-2-chloro-6-fluorobenzyloxybenzaldehyde

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-2-chloro-6-fluorobenzyloxybenzaldehyde
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart