APO455834262 Apollo Scientific Ltd

6-Fluoroisatoic anhydride

  • MDL number MFCD00112979
  • Cas Number : 78755-94-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 230
Synonym 5-Fluoro-2H-3,1-benzoxazine-2,4(1H)-dione, 5-Fluoro-1H-benzo[d][1,3]oxazine-2,4-dione
Density 1.502
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9104

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9104
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart