APO455834265 Apollo Scientific Ltd

3-Amino-3-(3-fluorophenyl)propanoic acid

  • MDL number MFCD00764457
  • Cas Number : 117391-51-2
  • Molecular Weight : 183.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 210-212
Synonym 3-Amino-3-(3-fluorophenyl)propionic acid, 3-(3-Fluorophenyl)-beta-alanine
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-3-3-fluorophenylpropanoic-acid

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-3-3-fluorophenylpropanoic-acid
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart