APO455834267 Apollo Scientific Ltd

7-Amino-2,4-bis(trifluoromethyl)-1,8-naphthyridine

  • MDL number MFCD00140205
  • Cas Number : 51420-72-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 195-197
Synonym 5,7-Bis(trifluoromethyl)-1,8-naphthyridin-2-amine
Purity 95
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9122

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9122
pictograms GHS07
hstatements H302;H312;H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart