APO455834271 Apollo Scientific Ltd

5-tert-Butyl-3-(trifluoromethyl)-1H-pyrazole

  • MDL number MFCD00729044
  • Cas Number : 150433-22-0
  • Molecular Weight : 192.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 178-180
Purity 97
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-tert-butyl-3-trifluoromethyl-1h-pyrazole

Safety Information

signalword Warning
pcodes P260;P280
MSDS https://store.apolloscientific.co.uk/product/5-tert-butyl-3-trifluoromethyl-1h-pyrazole
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart