APO455834274 Apollo Scientific Ltd

Pentafluorophenyl propane-2-sulphonate

  • MDL number MFCD05975149
  • Cas Number : 886361-27-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Perfluorophenyl propane-2-sulphonate
Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9174

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9174
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart