APO455834275 Apollo Scientific Ltd

Pentafluorophenyl 4-bromo-2-chlorobenzenesulphonate

  • MDL number MFCD05975147
  • Cas Number : 886361-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 88-89
Synonym Perfluorophenyl 4-bromo-2-chlorobenzenesulphonate
Purity 95
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9172

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9172
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart