APO455834281 Apollo Scientific Ltd

2-Bromo-6-fluorobenzenesulphonyl chloride

  • MDL number MFCD07368673
  • Cas Number : 886762-59-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29-30
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9182

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9182
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart