APO455834283 Apollo Scientific Ltd

2-Bromo-6-fluoroiodobenzene

  • MDL number MFCD07368675
  • Cas Number : 450412-29-0
  • Molecular Weight : 300.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-Bromo-3-fluoro-2-iodobenzene
Purity 99
Flash Point 104
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 108-109/10mm
Product Link https://store.apolloscientific.co.uk/product/2-bromo-6-fluoroiodobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-6-fluoroiodobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart