APO455834284 Apollo Scientific Ltd

4-Cyano-3-fluorobenzeneboronic acid

  • MDL number MFCD03411549
  • Cas Number : 843663-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 362-363
Synonym 4-Borono-2-fluorobenzonitrile
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9186

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9186
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart