APO455834289 Apollo Scientific Ltd

1,2-Dihydro-4-(4-fluorophenyl)-2-oxopyridine-3-carbonitrile

  • MDL number MFCD04117821
  • Cas Number : 886360-84-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 284-285
Synonym 3-Cyano-1,2-dihydro-4-(4-fluorophenyl)-2-oxopyridine, 1,2-Dihydro-4-(4-fluorophenyl)-2-oxonicotinonitrile
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9142

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9142
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart