APO455834291 Apollo Scientific Ltd

5-Fluoroisophthalonitrile

  • MDL number MFCD05663831
  • Cas Number : 453565-55-4
  • Molecular Weight : 146.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-111
Synonym 5-Fluorobenzene-1,3-dicarbonitrile
Purity 99
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-fluoroisophthalonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-fluoroisophthalonitrile
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart