APO455834292 Apollo Scientific Ltd

4-Hydroxy-3-(trifluoromethyl)benzonitrile

  • MDL number MFCD06797882
  • Cas Number : 124811-71-8
  • Molecular Weight : 187.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 171-175
Iupac Name 4-hydroxy-3-(trifluoromethyl)benzonitrile
Inchi Code 1S/C8H4F3NO/c9-8(10,11)6-3-5(4-12)1-2-7(6)13/h1-3,13H
Inchi Key QXIFYONUKBXFTH-UHFFFAOYSA-N

Description

Synonym 4-Cyano-2-(trifluoromethyl)phenol, 5-Cyano-2-hydroxybenzotrifluoride
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 171-175
Product Link https://store.apolloscientific.co.uk/product/4-hydroxy-3-trifluoromethylbenzonitrile

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/4-hydroxy-3-trifluoromethylbenzonitrile
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart