APO455834296 Apollo Scientific Ltd

2,5-Difluoro-4-nitrotoluene

  • MDL number MFCD06200905
  • Cas Number : 141412-60-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,5-Difluoro-4-methylnitrobenzene, 1,4-Difluoro-2-methyl-5-nitrobenzene
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/25-difluoro-4-nitrotoluene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/25-difluoro-4-nitrotoluene
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart