APO455834298 Apollo Scientific Ltd

2-Sulphanyl-4-(trifluoromethyl)pyridine

  • MDL number MFCD09881227
  • Cas Number : 121307-79-7
  • Molecular Weight : 179.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(trifluoromethyl)-2-pyridinethiol
Inchi Code 1S/C6H4F3NS/c7-6(8,9)4-1-2-10-5(11)3-4/h1-3H,(H,10,11)
Inchi Key SFLBZVBGZANGDN-UHFFFAOYSA-N

Description

Synonym 2-Mercapto-4-(trifluoromethyl)pyridine, 2-Sulphanyl-alpha,alpha,alpha-trifluoro-4-picoline, 4-(Trifluoromethyl)pyridine-2-thiol
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-sulphanyl-4-trifluoromethylpyridine

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-sulphanyl-4-trifluoromethylpyridine
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart