APO455834304 Apollo Scientific Ltd

1,1-Difluoro-2-iodoethane

  • MDL number MFCD04038311
  • Cas Number : 598-39-0
  • Molecular Weight : 191.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2,2-Difluoroethyl iodide
Purity 97
Density 2.18
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 86.8-87/740mm
Product Link https://store.apolloscientific.co.uk/product/11-difluoro-2-iodoethane

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/11-difluoro-2-iodoethane
pictograms GHS07
hstatements H302+H312+H332;H319
Availability details for
Quantity
  • Availability
  • Add to Cart