APO455834305 Apollo Scientific Ltd

3-Fluoro-2-methylbenzeneboronic acid

  • MDL number MFCD07368679
  • Cas Number : 163517-61-1
  • Molecular Weight : 153.95

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 142-144
Synonym 2-Borono-6-fluorotoluene
Purity 98
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-fluoro-2-methylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-fluoro-2-methylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart