APO455834310 Apollo Scientific Ltd

2-(Trifluoroacetyl)thiophene

  • MDL number MFCD00041427
  • Cas Number : 651-70-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(Thien-2-yl)-2,2,2-trifluoroethan-1-one
Purity 98
Density 1.403
Flash Point 59
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 163-165
Product Link https://store.apolloscientific.co.uk/product/2-trifluoroacetylthiophene

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-trifluoroacetylthiophene
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart