APO455834313 Apollo Scientific Ltd

4-Bromo-3-fluoro-2-nitroaniline

  • MDL number MFCD07368682
  • Cas Number : 886762-75-0
  • Molecular Weight : 235.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 6-Amino-3-bromo-2-fluoronitrobenzene
Purity 97
Flash Point >110
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-3-fluoro-2-nitroaniline

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/4-bromo-3-fluoro-2-nitroaniline
pictograms GHS07
hstatements H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart