APO455834318 Apollo Scientific Ltd

3'-Amino-4'-fluoroacetanilide

  • MDL number MFCD07368688
  • Cas Number : 113596-04-6
  • Molecular Weight : 168.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(3-amino-4-fluorophenyl)acetamide
Inchi Code 1S/C8H9FN2O/c1-5(12)11-6-2-3-7(9)8(10)4-6/h2-4H,10H2,1H3,(H,11,12)
Inchi Key RSZIKGBUIWSQSA-UHFFFAOYSA-N

Description

Synonym N-(3-Amino-4-fluorophenyl)acetamide, 5-(Acetylamino)-2-fluoroaniline
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-amino-4-fluoroacetanilide

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-amino-4-fluoroacetanilide
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart