APO455834328 Apollo Scientific Ltd

2,3,6-Trifluorophenacyl bromide

  • MDL number MFCD07368691
  • Cas Number : 886762-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 29.8-30-3
Synonym 2-Bromo-2',3',6'-trifluoroacetophenone, 2-Bromo-1-(2,3,6-trifluorophenyl)ethan-1-one
Storage temp. Ambient Temperature
Physical Form Solid-Low Melt
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9249

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9249
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart