APO455834329 Apollo Scientific Ltd

2-Chloro-6-fluorophenacyl bromide

  • MDL number MFCD07368692
  • Cas Number : 157359-99-4
  • Molecular Weight : 251.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Bromo-2'-chloro-6'-fluoroacetophenone, 2-Bromo-1-(2-chloro-6-fluorophenyl)ethan-1-one
Purity 98
Physical Form Liquid-Clear
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 74/0.06mm
Product Link https://store.apolloscientific.co.uk/product/2-chloro-6-fluorophenacyl-bromide

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-6-fluorophenacyl-bromide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart