APO455834333 Apollo Scientific Ltd

2',6'-Dichloro-4'-fluoroacetanilide

  • MDL number MFCD07368695
  • Cas Number : 392-16-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 186-187
Synonym N-(2,6-Dichloro-4-fluorophenyl)acetamide
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9277

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9277
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart