APO455834334 Apollo Scientific Ltd

3-(Methylamino)benzotrifluoride

  • MDL number MFCD07368696
  • Cas Number : 2026-70-2
  • Molecular Weight : 175.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-methyl-3-(trifluoromethyl)aniline
Inchi Code 1S/C8H8F3N/c1-12-7-4-2-3-6(5-7)8(9,10)11/h2-5,12H,1H3
Inchi Key SRTKIHVQZYXHHJ-UHFFFAOYSA-N

Description

Synonym N-Methyl-3-(trifluoromethyl)aniline, N-Methyl-alpha,alpha,alpha-trifluoro-m-toluidine
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-methylaminobenzotrifluoride

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/3-methylaminobenzotrifluoride
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart