APO455834335 Apollo Scientific Ltd

2-Fluoro-6-hydroxybenzonitrile

  • MDL number MFCD03428592
  • Cas Number : 140675-43-0
  • Molecular Weight : 137.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 154-158
Synonym 2-Cyano-3-fluorophenol, 6-Fluorosalicylonitrile
Purity 99
Flash Point >110
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-6-hydroxybenzonitrile

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-6-hydroxybenzonitrile
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart