APO455834338 Apollo Scientific Ltd

1,4-Dimethyl-2-fluorobenzene

  • MDL number MFCD03412243
  • Cas Number : 696-01-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point -6
Synonym 2-Fluoro-p-xylene
Flash Point 39
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 137-138
Product Link https://store.apolloscientific.co.uk/product/14-dimethyl-2-fluorobenzene

Safety Information

signalword Warning
pcodes P210;P280
MSDS https://store.apolloscientific.co.uk/product/14-dimethyl-2-fluorobenzene
pictograms GHS02;GHS07
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart