APO455834344 Apollo Scientific Ltd

N-[(2,4-Difluorophenyl)carbamothioyl]benzamide

  • MDL number MFCD07368702
  • Cas Number : 887266-92-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 117-120
Synonym 1-Benzoyl-3-(2,4-difluorophenyl)thiourea
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9464

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9464
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart