APO455834348 Apollo Scientific Ltd

2-Methoxy-5-(trifluoromethoxy)benzaldehyde

  • MDL number MFCD04115958
  • Cas Number : 145742-65-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 5-(Trifluoromethoxy)-o-anisaldehyde
Density 1.36
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9474

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9474
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart