APO455834354 Apollo Scientific Ltd

5-Bromo-2-fluoro-4-methylaniline

  • MDL number MFCD11504886
  • Cas Number : 945244-29-1
  • Molecular Weight : 204.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 4-Amino-2-bromo-5-fluorotoluene, 5-Bromo-2-fluoro-p-toluidine
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-fluoro-4-methylaniline

Safety Information

signalword Warning
pcodes P260;P280;P312
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-fluoro-4-methylaniline
pictograms GHS07
hstatements H302+H312+H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart