APO455834358 Apollo Scientific Ltd

2,6-Bis(trifluoromethyl)benzeneboronic acid

  • MDL number MFCD04039321
  • Cas Number : 681812-07-7
  • Molecular Weight : 257.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point ca 150
Storage temp. Store at -20
Physical Form Solid-Powder
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/26-bistrifluoromethylbenzeneboronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/26-bistrifluoromethylbenzeneboronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart