APO455834361 Apollo Scientific Ltd

5-Bromo-2-fluoropyridine-3-boronic acid

  • MDL number MFCD04039317
  • Cas Number : 501435-91-2
  • Molecular Weight : 219.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-bromo-2-fluoropyridine-3-boronic-acid

Safety Information

signalword Warning
pcodes P280;P261;P271
MSDS https://store.apolloscientific.co.uk/product/5-bromo-2-fluoropyridine-3-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart