APO455834366 Apollo Scientific Ltd

3,5-Dichloro-2-fluoropyridine

  • MDL number MFCD04039349
  • Cas Number : 823-56-3
  • Molecular Weight : 165.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 41-44
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/35-dichloro-2-fluoropyridine

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/35-dichloro-2-fluoropyridine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart