APO455834372 Apollo Scientific Ltd

2,3,5-Trifluoropyridine

  • MDL number MFCD03001162
  • Cas Number : 76469-41-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Density 1.4
Flash Point 30
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 102
Product Link https://store.apolloscientific.co.uk/product/235-trifluoropyridine

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/235-trifluoropyridine
pictograms GHS02
hstatements H226;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart