APO455834373 Apollo Scientific Ltd

2-Fluoro-6-nitroaniline

  • MDL number MFCD04112583
  • Cas Number : 17809-36-8
  • Molecular Weight : 156.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 74-75
Iupac Name 2-fluoro-6-nitroaniline
Inchi Code 1S/C6H5FN2O2/c7-4-2-1-3-5(6(4)8)9(10)11/h1-3H,8H2
Inchi Key BHWHYGWMNMCXBA-UHFFFAOYSA-N

Description

Synonym 2-Amino-3-fluoronitrobenzene
Purity 97%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 74-75
Product Link https://store.apolloscientific.co.uk/product/2-fluoro-6-nitroaniline

Safety Information

signalword Warning
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/2-fluoro-6-nitroaniline
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart