APO455834378 Apollo Scientific Ltd

Pentafluorophenyl methanesulphonate

  • MDL number MFCD06659544
  • Cas Number : 161912-36-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Pentafluorophenyl mesylate, Perfluorophenyl methanesulphonate
Flash Point >110
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Boiling point 76/0.6mm
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9312

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9312
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart