APO455834380 Apollo Scientific Ltd

3,5-Difluorophenylthiourea

  • MDL number MFCD07368707
  • Cas Number : 791594-33-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 146-148
Synonym 1-(3,5-Difluorophenyl)thiourea, 1,3-Difluoro-5-thioureidobenzene
Storage temp. Ambient Temperature
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9378

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9378
pictograms GHS06
hstatements H301;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart