APO455834383 Apollo Scientific Ltd

4-(4-Fluorobenzyloxy)cinnamic acid

  • MDL number MFCD00173647
  • Cas Number : 175136-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 209-210
Synonym trans-3-[4-(4-Fluorobenzyloxy)phenyl]acrylic acid
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9389

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9389
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart