APO455834384 Apollo Scientific Ltd

N-(2,4-Difluorophenyl)glycinehydrazide

  • MDL number MFCD00098762
  • Cas Number : 2351-00-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-(2,4-Difluoroanilino)acetohydrazide, 2-[(2,4-Difluorophenyl)amino]acetohydrazide
Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9394

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9394
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart