APO455834397 Apollo Scientific Ltd

2-Methyl-4-(trifluoromethoxy)benzaldehyde

  • MDL number MFCD07368717
  • Cas Number : 886763-07-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 2-Formyl-5-(trifluoromethoxy)toluene, 4-(Trifluoromethoxy)-o-tolualdehyde
Purity 98
Storage temp. Ambient Temperature
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=9399

Safety Information

signalword Warning
pcodes P271;P280;P261
MSDS http://www.apolloscientific.co.uk/display_item.php?id=9399
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart