APO455834399 Apollo Scientific Ltd

3,3,3-Trifluoropropanoyl chloride

  • MDL number MFCD00800953
  • Cas Number : 41463-83-6
  • Molecular Weight : 146.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3,3,3-Trifluoropropionyl chloride
Density 1.422
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 71-72
Product Link https://store.apolloscientific.co.uk/product/333-trifluoropropanoyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/333-trifluoropropanoyl-chloride
pictograms GHS05;GHS07;GHS09
hstatements H314;H335;EUH059
Availability details for
Quantity
  • Availability
  • Add to Cart