APO455834414 Apollo Scientific Ltd

2-(Aminomethyl)-3-fluoro-6-methylpyridine hydrochloride

  • MDL number MFCD17214195
  • Cas Number : 1257535-11-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym (3-Fluoro-6-methylpyridin-2-yl)methylamine hydrochloride
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=48226

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=48226
pictograms GHS07
hstatements H302;H312;H332;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart